4-Benzyloxyindole ≥98.0%
Supplier: TCI America
B2811-5G
B2811-1G
TCB2811-5GEA
371.3
CAD
TCB2811-5G
TCB2811-1G
4-Benzyloxyindole ≥98.0%
4-Benzyloxyindole
CAS Number: 20289-26-3
Molecular Formula: C15H13NO
Molecular Weight: 223.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62
Formula:
C₁₅H₁₃NO MW: 223.27 g/mol Melting Pt: 62 °C |
MDL Number:
MFCD00047200 CAS Number: 20289-26-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...