6-(Benzyloxy)-1H-indole ≥98.0%

Supplier: TCI America

B2809-25G B2809-5G B2809-1G
TCB2809-25GEA 1213.44 CAD
TCB2809-25G TCB2809-5G TCB2809-1G
6-(Benzyloxy)-1H-indole ≥98.0%
6-(Benzyloxy)-1H-indole

CAS Number: 15903-94-3
Molecular Formula: C15H13NO
Molecular Weight: 223.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117

Formula: C15H13NO
Melting Pt: 117 °C
MDL Number: MFCD00053554
CAS Number: 15903-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7fcc013585b62d33d3d4c05fd6d60234859e93c&vid=f7fcc013585b62d33d3d4c05fd6d60234859e93c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR