2-Bromo-5-fluoropyridine ≥98.0%
Supplier: TCI America
B2799-25G
B2799-5G
TCB2799-25GEA
290.88
CAD
TCB2799-25G
TCB2799-5G
2-Bromo-5-fluoropyridine ≥98.0%
2-Bromo-5-fluoropyridine
CAS Number: 41404-58-4
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 83
Flash Point (°C): 73
Freezing point (°C): 29
Formula:
C₅H₃BrFN Boiling Pt: 83 °C Flash Pt: 73 °C |
MDL Number:
MFCD00234011 CAS Number: 41404-58-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...