4-Benzoylbutyric acid ≥96.0% (by GC, titration analysis)
Supplier: TCI America
B2768-5G
B2768-25G
TCB2768-5GEA
66.36
CAD
TCB2768-5G
TCB2768-25G
4-Benzoylbutyric acid ≥96.0% (by GC, titration analysis)
4-Benzoylbutyric acid
CAS Number: 1501-05-9
MDL Number: MFCD00004411
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 129
Other Product Names / Synonyms: 5-Oxo-5-phenylvaleric Acid, 5-Oxo-5-phenylpentanoic Acid
Formula:
C₁₁H₁₂O₃ Melting Pt: 129 °C |
MDL Number:
MFCD00004411 CAS Number: 1501-05-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...