5-Bromo-2-nitropyridine ≥98.0%
Supplier: TCI America
B2766-5G
B2766-1G
TCB2766-5GEA
91.64
CAD
TCB2766-5G
TCB2766-1G
5-Bromo-2-nitropyridine ≥98.0%
5-Bromo-2-nitropyridine
CAS Number: 39856-50-3
Molecular Formula: C5H3BrN2O2
Molecular Weight: 203.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150
Formula:
C₅H₃BrN₂O₂ Melting Pt: 150 °C |
MDL Number:
MFCD00160411 CAS Number: 39856-50-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...