2-Bromo-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2722-5G B2722-25G
TCB2722-5GEA 193.39 CAD
TCB2722-5G TCB2722-25G
2-Bromo-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-5-fluorobenzoic acid

CAS Number: 394-28-5
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00142874
CAS Number: 394-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e168426facbadd284ee9847e98f7ff1789bfc9b5&vid=e168426facbadd284ee9847e98f7ff1789bfc9b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR