4,6-Bis(diphenylphosphino)phenoxazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4,6-Bis(diphenylphosphino)-10H-phenoxazine, NIXANTPHOS

B2717-100MG B2717-1G
TCB2717-100MGEA 71.1 CAD
TCB2717-100MG TCB2717-1G
4,6-Bis(diphenylphosphino)phenoxazine ≥98.0% (by HPLC, total nitrogen)
4,6-Bis(diphenylphosphino)phenoxazine

CAS Number: 261733-18-0
MDL Number: MFCD03788937
Molecular Formula: C36H27NOP2
Molecular Weight: 551.57
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₃₆H₂₇NOP₂
MW: 551.57 g/mol
MDL Number: MFCD03788937
CAS Number: 261733-18-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12ce5994c8c007f629d2f8a3d6da00b789aa1dd2&vid=12ce5994c8c007f629d2f8a3d6da00b789aa1dd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR