1,1'-Bis(di-tert-butylphosphino)ferrocene ≥95.0% (by titrimetric analysis)
Supplier: TCI America
B2711-100MG
B2711-1G
TCB2711-100MGEA
37.92
CAD
TCB2711-100MG
TCB2711-1G
1,1'-Bis(di-tert-butylphosphino)ferrocene ≥95.0% (by titrimetric analysis)
1,1'-Bis(di-tert-butylphosphino)ferrocene
CAS Number: 84680-95-5
Molecular Formula: C26H44FeP2
Molecular Weight: 474.43
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 182
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Bis[(di-tert-butylphosphino)cyclopentadienyl]iron, dtbpf
Formula:
C₂₆H₄₄FeP₂ Melting Pt: 182 °C |
MDL Number:
MFCD01630818 CAS Number: 84680-95-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...