1,1'-Bis(diisopropylphosphino)ferrocene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene
B2710-100MG
B2710-1G
TCB2710-100MGEA
61.62
CAD
TCB2710-100MG
TCB2710-1G
1,1'-Bis(diisopropylphosphino)ferrocene ≥98.0% (by titrimetric analysis)
1,1'-Bis(diisopropylphosphino)ferrocene
CAS Number: 97239-80-0
Molecular Formula: C22H36FeP2
Molecular Weight: 418.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 50
Flash Point (°C): 110
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands
Other Product Names / Synonyms: Bis[(diisopropylphosphino)cyclopentadienyl]iron
Formula:
C₂₂H₂₈FeP₂ MW: 410.25 g/mol Melting Pt: 50 °C Flash Pt: 110 °C |
MDL Number:
MFCD02684559 CAS Number: 97239-80-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...