(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0% (by HPLC)
Supplier: TCI America
B2709-1G
B2709-5G
B2709-25G
TCB2709-1GEA
90.19
CAD
TCB2709-1G
TCB2709-5G
TCB2709-25G
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥98.0% (by HPLC)
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
CAS Number: 161265-03-8
Molecular Formula: C39H32OP2
Molecular Weight: 578.63
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 229
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands, Ligands ([2+2+2]Cycloaddition Reaction), Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Xantphos
Formula:
C₃₉H₃₂OP₂ Melting Pt: 229 °C |
MDL Number:
MFCD00233866 CAS Number: 161265-03-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...