5-Bromo-2-hydroxy-3-nitropyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one

B2706-1G B2706-5G
TCB2706-1GEA 44.24 CAD
TCB2706-1G TCB2706-5G
5-Bromo-2-hydroxy-3-nitropyridine ≥98.0%
5-Bromo-2-hydroxy-3-nitropyridine

CAS Number: 15862-34-7
Molecular Formula: C5H3BrN2O3
Molecular Weight: 218.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 247


Other Product Names / Synonyms: 5-Bromo-3-nitro-2-pyridinol

Formula: C₅H₃BrN₂O₃
MW: 218.99 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00023473
CAS Number: 15862-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f2cc0ff960ac4c5c58e6bfcf66589322760fb3c&vid=5f2cc0ff960ac4c5c58e6bfcf66589322760fb3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR