5-Bromo-2-fluoropyridine ≥98.0%
Supplier: TCI America
B2705-25G
B2705-5G
TCB2705-25GEA
409.85
CAD
TCB2705-25G
TCB2705-5G
5-Bromo-2-fluoropyridine ≥98.0%
5-Bromo-2-fluoropyridine
CAS Number: 766-11-0
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): 73
Specific Gravity (20/20): 1.72
Formula:
C₅H₃BrFN Boiling Pt: 63 °C Density: 1.72 (20 °C) Flash Pt: 73 °C |
MDL Number:
MFCD01863742 CAS Number: 766-11-0 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...