(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥93.0%
Supplier: TCI America
B2704-100MG
B2704-1G
TCB2704-100MGEA
88.48
CAD
TCB2704-100MG
TCB2704-1G
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane ≥93.0%
(S,S)-(-)-2,3-Bis(diphenylphosphino)butane
CAS Number: 64896-28-2
Molecular Formula: C28H28P2
Molecular Weight: 426.48
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 109
Specific rotation [a]20/D: -180 deg (C=1.5, CHCl3)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-Chiraphos
Formula:
C₂₈H₂₈P₂ Melting Pt: 109 °C |
MDL Number:
MFCD00009836 CAS Number: 64896-28-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...