1-Butyl-4-methylpyridiniumhexafluorophosphate ≥97.0%
Supplier: TCI America
Synonyms:
1-Butyl-4-methylpyridinium Hexafluorophosphate
B2701-25G
B2701-5G
TCB2701-25GEA
338.91
CAD
TCB2701-25G
TCB2701-5G
1-Butyl-4-methylpyridiniumhexafluorophosphate ≥97.0%
1-Butyl-4-methylpyridinium hexafluorophosphate
CAS Number: 401788-99-6
Molecular Formula: C10H16F6NP
Molecular Weight: 295.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 42
Formula:
C₁₀H₁₆F₆NP MW: 295.21 g/mol Melting Pt: 42 °C |
MDL Number:
MFCD03095438 CAS Number: 401788-99-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...