2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99.0%, purified by sublimation

Supplier: TCI America

Synonyms: PBD, 2-([1,1-Biphenyl]-4-yl)-5-(4-(tert-butyl)phenyl)-1,3,4-oxadiazole, 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole

B2696-1G B2696-5G
TCB2696-1GEA 161.95 CAD
TCB2696-1G TCB2696-5G
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole ≥99.0%, purified by sublimation
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole

CAS Number: 15082-28-7
MDL Number: MFCD00003101
Molecular Formula: C24H22N2O
Molecular Weight: 354.45
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 138


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Highly Purified Reagents
Other Product Names / Synonyms: 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole, PBD

Formula: C₂₄H₂₂N₂O
MW: 354.45 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00003101
CAS Number: 15082-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a3db799f4565f91bdc5ee719108e672d43ee6dc&vid=aac4e45748a8d834586027762ea61ecb8fbcc406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR