1-Bromo-4-cyclohexylbenzene ≥97.0%

Supplier: TCI America

B2686-25G B2686-5G
TCB2686-25GEA 584.6 CAD
TCB2686-25G TCB2686-5G
1-Bromo-4-cyclohexylbenzene ≥97.0%
1-Bromo-4-cyclohexylbenzene

CAS Number: 25109-28-8
Molecular Formula: C12H15Br
Molecular Weight: 239.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.29


Other Product Names / Synonyms: 4-Bromophenylcyclohexane

Formula: C₁₂H₁₅Br
Density: 1.29 (20 °C)
MDL Number: MFCD00019339
CAS Number: 25109-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0218719730e4f43c2feb66b70878254e6c368d0&vid=508efaf9632d4a97586da5d05fdcf93a0f2da3d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR