Bis(4-fluorophenyl)ether ≥98.0%

Supplier: TCI America

B2648-1G B2648-5G
TCB2648-1GEA 37.92 CAD
TCB2648-1G TCB2648-5G
Bis(4-fluorophenyl)ether ≥98.0%
Bis(4-fluorophenyl)ether

CAS Number: 330-93-8
Molecular Formula: C12H8F2O
Molecular Weight: 206.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 110
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4,4'-Difluorodiphenyl Ether

Formula: C₁₂H₈F₂O
Boiling Pt: 240 °C
Density: 1.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013552
CAS Number: 330-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a572f5a8c02386451ae4d993efd9b561ed143d&vid=88a572f5a8c02386451ae4d993efd9b561ed143d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR