2,2'-Bis(diphenylphosphino)biphenyl ≥98.0%

Supplier: TCI America

B2630-100MG B2630-1G
TCB2630-100MGEA 47.4 CAD
TCB2630-100MG TCB2630-1G
2,2'-Bis(diphenylphosphino)biphenyl ≥98.0%
2,2'-Bis(diphenylphosphino)biphenyl

CAS Number: 84783-64-2
Molecular Formula: C36H28P2
Molecular Weight: 522.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 215

Formula: C₃₆H₂₈P₂
Melting Pt: 215 °C
MDL Number: MFCD03094574
CAS Number: 84783-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=805af03d0d00705af8dda9a93f9ad5984b103e0c&vid=805af03d0d00705af8dda9a93f9ad5984b103e0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR