4-Butylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-n-Butylbenzeneboronic acid
B2589-1G
B2589-5G
TCB2589-1GEA
50.56
CAD
TCB2589-1G
TCB2589-5G
4-Butylphenylboronic acid (contains varying amounts of Anhydride)
4-Butylphenylboronic acid
CAS Number: 145240-28-4
MDL Number: MFCD02093926
Molecular Formula: C10H15BO2
Molecular Weight: 178.04
Form: Crystal
Color: White
Melting point (°C): 97
Other Product Names / Synonyms: 4-Butylbenzeneboronic Acid
Formula:
C₁₀H₁₅BO₂ MW: 178.04 g/mol Melting Pt: 97 °C |
MDL Number:
MFCD02093926 CAS Number: 145240-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...