5-(2-Bromophenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-(2-Bromophenyl)tetrazole

B2577-25G B2577-5G
TCB2577-25GEA 229.73 CAD
TCB2577-25G TCB2577-5G
5-(2-Bromophenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-(2-Bromophenyl)-1H-tetrazole

CAS Number: 73096-42-1
MDL Number: MFCD00151799
Molecular Formula: C7H5BrN4
Molecular Weight: 225.05
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₇H₅BrN₄
MW: 225.05 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00151799
CAS Number: 73096-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80bb7015d0217a54ef84e086e69848fd6e2196ab&vid=80bb7015d0217a54ef84e086e69848fd6e2196ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR