5-Bromo-2-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-picoline, 3-Bromo-6-picoline

B2572-1G B2572-5G
TCB2572-1GEA 31.6 CAD
TCB2572-1G TCB2572-5G
5-Bromo-2-methylpyridine ≥98.0%
5-Bromo-2-methylpyridine

CAS Number: 3430-13-5
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 74
Melting point (°C): 36


Other Product Names / Synonyms: 5-Bromo-2-picoline

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 74 °C
Melting Pt: 36 °C
MDL Number: MFCD00661170
CAS Number: 3430-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f6ef7688ba6a9e8ab24155cb9673cf564a5241&vid=82f6ef7688ba6a9e8ab24155cb9673cf564a5241&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR