1-Bromo-2,5-bis(trifluoromethyl)benzene ≥87.0%

Supplier: TCI America

Synonyms: 2,5-Bis(trifluoromethyl)bromobenzene, 2-Bromo-1,4-bis(trifluoromethyl)benzene

B2545-5G B2545-25G
TCB2545-5GEA 47.87 CAD
TCB2545-5G TCB2545-25G
1-Bromo-2,5-bis(trifluoromethyl)benzene ≥87.0%
1-Bromo-2,5-bis(trifluoromethyl)benzene

CAS Number: 7617-93-8
MDL Number: MFCD03094336
Molecular Formula: C8H3BrF6
Molecular Weight: 293.01
Purity/Analysis Method: >87.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Melting point (°C): 5
Flash Point (°C): 110
Specific Gravity (20/20): 1.75

Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 158 °C
Melting Pt: 5 °C
Density: 1.75 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD03094336
CAS Number: 7617-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e393aadde19532c02bdbd0c57638046d53dd26e&vid=4e393aadde19532c02bdbd0c57638046d53dd26e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 87.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR