α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥98.0%
Supplier: TCI America
B2544-25G
B2544-5G
TCB2544-25GEA
295.93
CAD
TCB2544-25G
TCB2544-5G
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥98.0%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
CAS Number: 328-93-8
MDL Number: MFCD00074940
Molecular Formula: C8H5F6N
Molecular Weight: 229.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 71
Specific Gravity (20/20): 1.48
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-2,5-xylidine
Formula:
C₈H₅F₆N Boiling Pt: 177 °C Density: 1.48 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00074940 CAS Number: 328-93-8 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...