4,4'-Biphenyldiboronic acid ≥95.0%
Supplier: TCI America
Synonyms:
[1,1'-Biphenyl]-4,4'-diyldiboronic acid
B2490-1G
B2490-5G
TCB2490-1GEA
178.54
CAD
TCB2490-1G
TCB2490-5G
4,4'-Biphenyldiboronic acid ≥95.0%
4,4'-Biphenyldiboronic acid
CAS Number: 4151-80-8
Molecular Formula: C12H12B2O4
Molecular Weight: 241.84
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 300
Formula:
C₁₂H₁₂B₂O₄ MW: 241.85 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00151795 CAS Number: 4151-80-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...