3-Biphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

B2489-5G B2489-25G
TCB2489-5GEA 153.26 CAD
TCB2489-5G TCB2489-25G
3-Biphenylboronic acid (contains varying amounts of Anhydride)
3-Biphenylboronic acid

CAS Number: 5122-95-2
MDL Number: MFCD01318102
Molecular Formula: C12H11BO2
Molecular Weight: 198.03
Form: Crystal
Color: White
Melting point (°C): 208

Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
Melting Pt: 208 °C
MDL Number: MFCD01318102
CAS Number: 5122-95-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8d8bf61200867b40b4b1fb6fcd9dfea11879164&vid=b8d8bf61200867b40b4b1fb6fcd9dfea11879164&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR