1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI America
B2474-25G
B2474-5G
TCB2474-25GEA
510.7
CAD
TCB2474-25G
TCB2474-5G
1-Butyl-2,3-dimethylimidazoliumhexafluorophosphate ≥97.0% (by total nitrogen and titration analysis)
1-Butyl-2,3-dimethylimidazolium hexafluorophosphate
CAS Number: 227617-70-1
Molecular Formula: C9H17F6N2P
Molecular Weight: 298.21
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal
Melting point (°C): 38
Formula:
C₉H₁₇F₆N₂P Melting Pt: 38 °C |
MDL Number:
MFCD03790877 CAS Number: 227617-70-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...