1-Butyl-2,3-dimethylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2473-5G B2473-25G
TCB2473-5GEA 64.78 CAD
TCB2473-5G TCB2473-25G
1-Butyl-2,3-dimethylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Butyl-2,3-dimethylimidazolium chloride

CAS Number: 98892-75-2
Molecular Formula: C9H17ClN2
Molecular Weight: 188.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 99

Formula: C₉H₁₇ClN₂
Melting Pt: 99 °C
MDL Number: MFCD03093291
CAS Number: 98892-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b96e991a036ebe2b44386594364bacebad5dd3f1&vid=b96e991a036ebe2b44386594364bacebad5dd3f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR