1-Boc-piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2415-5G B2415-25G
TCB2415-5GEA 113.76 CAD
TCB2415-5G TCB2415-25G
1-Boc-piperazine ≥98.0% (by GC, titration analysis)
1-Boc-piperazine

CAS Number: 57260-71-6
MDL Number: MFCD00075265
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110


Other Product Names / Synonyms: 1-Boc-piperazine, tert-Butyl Piperazine-1-carboxylate

Formula: C₉H₁₈N₂O₂
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00075265
CAS Number: 57260-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ea20ecd1b7d405cb0480ae02eb9549107cb65cf&vid=3ea20ecd1b7d405cb0480ae02eb9549107cb65cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR