4-Methoxy-1-butanol ≥98.0%
Supplier: TCI America
Synonyms:
4-Methoxybutan-1-ol
B2402-25G
B2402-5G
TCB2402-25GEA
360.24
CAD
TCB2402-25G
TCB2402-5G
4-Methoxy-1-butanol ≥98.0%
4-Methoxy-1-butanol
CAS Number: 111-32-0
MDL Number: MFCD00040437
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Specific Gravity (20/20): 0.93
Other Product Names / Synonyms: 4-Methoxy-1-butanol, Tetramethylene Glycol Monomethyl Ether
Formula:
C₅H₁₂O₂ MW: 104.15 Boiling Pt: 66 °C Density: 0.93 (20 °C) |
MDL Number:
MFCD00040437 CAS Number: 111-32-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...