5-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2398-25G B2398-5G
TCB2398-25GEA 134.3 CAD
TCB2398-25G TCB2398-5G
5-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-methoxypyridine

CAS Number: 13472-85-0
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 96
Specific Gravity (20/20): 1.56

Formula: C₆H₆BrNO
MW: 188.02
Boiling Pt: 195 °C
Density: 1.56 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD01318952
CAS Number: 13472-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce9439412a7247517562cac4d7c39ac1683519b6&vid=ce9439412a7247517562cac4d7c39ac1683519b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR