3,5-Bis(trifluoromethyl)thiophenol ≥96.0% (by GC, titration analysis)

Supplier: TCI America

B2391-1G B2391-5G
TCB2391-1GEA 116.92 CAD
TCB2391-1G TCB2391-5G
3,5-Bis(trifluoromethyl)thiophenol ≥96.0% (by GC, titration analysis)
3,5-Bis(trifluoromethyl)thiophenol

CAS Number: 130783-02-7
MDL Number: MFCD00042273
Molecular Formula: C8H4F6S
Molecular Weight: 246.17
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 167
Flash Point (°C): 65


Product Categories: Synthetic Organic Chemistry, Thiols
Other Product Names / Synonyms: 3,5-Bis(trifluoromethyl)thiophenol

Formula: C₈H₄F₆S
Boiling Pt: 167 °C
Flash Pt: 65 °C
MDL Number: MFCD00042273
CAS Number: 130783-02-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=304cf4cee122fb46abc9844abb37b2f153d1a963&vid=304cf4cee122fb46abc9844abb37b2f153d1a963&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR