4-tert-Butoxyphenol ≥98.0%
Supplier: TCI America
Synonyms:
Hydroquinone Mono-tert-butyl Ether
B2390-1G
B2390-5G
TCB2390-1GEA
84.37
CAD
TCB2390-1G
TCB2390-5G
4-tert-Butoxyphenol ≥98.0%
4-tert-Butoxyphenol
CAS Number: 2460-87-9
MDL Number: MFCD02183556
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: Hydroquinone Mono-tert-butyl Ether
Formula:
C₁₀H₁₄O₂ Melting Pt: 153 °C |
MDL Number:
MFCD02183556 CAS Number: 2460-87-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...