2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97.0%

Supplier: TCI America

Synonyms: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP

B2383-25G B2383-5G
TCB2383-25GEA 677.82 CAD
TCB2383-25G TCB2383-5G
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥97.0%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

CAS Number: 98327-87-8
MDL Number: MFCD00010805
Molecular Formula: C44H32P2
Molecular Weight: 622.69
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 282


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (+/-)-BINAP

Formula: C₄₄H₃₂P₂
MW: 622.67 g/mol
Melting Pt: 282 °C
MDL Number: MFCD00010805
CAS Number: 98327-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a98608e38c2331cd9df715c9236f9c9b853ec5&vid=39a98608e38c2331cd9df715c9236f9c9b853ec5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR