2-Bromo-5-fluorobenzaldehyde ≥95.0%

Supplier: TCI America

B2353-25G B2353-5G
TCB2353-25GEA 246.32 CAD
TCB2353-25G TCB2353-5G
2-Bromo-5-fluorobenzaldehyde ≥95.0%
2-Bromo-5-fluorobenzaldehyde

CAS Number: 94569-84-3
MDL Number: MFCD00142872
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 55
Flash Point (°C): 87

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Melting Pt: 55 °C
Flash Pt: 87 °C
MDL Number: MFCD00142872
CAS Number: 94569-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b55991607aebee2b41b2e17844b100db439b2f83&vid=b55991607aebee2b41b2e17844b100db439b2f83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR