3-Bromothioanisole ≥98.0%

Supplier: TCI America

B2346-25G B2346-5G
TCB2346-25GEA 443.98 CAD
TCB2346-25G TCB2346-5G
3-Bromothioanisole ≥98.0%
3-Bromothioanisole

CAS Number: 33733-73-2
MDL Number: MFCD00041395
Molecular Formula: C7H7BrS
Molecular Weight: 203.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 250
Flash Point (°C): 110
Specific Gravity (20/20): 1.52


Other Product Names / Synonyms: 1-Bromo-3-(methylthio)benzene, 3-Bromophenyl Methyl Sulfide

Formula: C₇H₇BrS
Boiling Pt: 250 °C
Density: 1.52 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041395
CAS Number: 33733-73-2
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR