5-Bromo-1-indanone ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one

B2342-5G B2342-25G B2342-1G
TCB2342-5GEA 122.18 CAD
TCB2342-5G TCB2342-25G TCB2342-1G
5-Bromo-1-indanone ≥98.0%
5-Bromo-1-indanone

CAS Number: 34598-49-7
MDL Number: MFCD00082718
Molecular Formula: C9H7BrO
Molecular Weight: 211.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127

Formula: C₉H₇BrO
MW: 211.06 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00082718
CAS Number: 34598-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d06c1e77687edccc75147cde3f3b3ba8bceb238&vid=4d06c1e77687edccc75147cde3f3b3ba8bceb238&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR