1-Bromo-4-fluoro-2-nitrobenzene ≥97.0%
Supplier: TCI America
B2339-1G
B2339-5G
TCB2339-1GEA
41.08
CAD
TCB2339-1G
TCB2339-5G
1-Bromo-4-fluoro-2-nitrobenzene ≥97.0%
1-Bromo-4-fluoro-2-nitrobenzene
CAS Number: 446-09-3
MDL Number: MFCD00055530
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220.00
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 41
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-5-fluoronitrobenzene
Formula:
C₆H₃BrFNO₂ Boiling Pt: 150 °C Melting Pt: 41 °C |
MDL Number:
MFCD00055530 CAS Number: 446-09-3 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...