1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
1-Butyl-3-methylimidazolium Trifluoromethanesulfonate, 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate
B2337-25G
B2337-5G
TCB2337-25GEA
561.11
CAD
TCB2337-25G
TCB2337-5G
1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate ≥98.0%
1-Butyl-3-methylimidazolium trifluoromethanesulphonate
CAS Number: 174899-66-2
MDL Number: MFCD03427620
Molecular Formula: C9H15F3N2O3S
Molecular Weight: 288.29
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Melting point (°C): 16
Specific Gravity (20/20): 1.31
Formula:
C₉H₁₅F₃N₂O₃S MW: 288.29 g/mol Melting Pt: 16 °C Density: 1.31 (20 °C) |
MDL Number:
MFCD03427620 CAS Number: 174899-66-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...