3-Bromo-2-hydroxypyridine ≥97.0%

Supplier: TCI America

Synonyms: 3-Bromopyridin-2(1H)-one, 3-Bromopyridin-2-ol, 3-Bromo-2-pyridinol

B2330-25G B2330-5G
TCB2330-25GEA 418.7 CAD
TCB2330-25G TCB2330-5G
3-Bromo-2-hydroxypyridine ≥97.0%
3-Bromo-2-hydroxypyridine

CAS Number: 13466-43-8
MDL Number: MFCD03411569
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 3-Bromo-2-pyridinol, 3-Bromo-2(1H)-pyridinone, 3-Bromo-2-pyridone

Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 182 °C
MDL Number: MFCD03411569
CAS Number: 13466-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=054b0ef6bad031ea9f433d1ab6b7be964a2fb971&vid=054b0ef6bad031ea9f433d1ab6b7be964a2fb971&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR