5-Bromo-2-chloropyridine ≥98.0%
Supplier: TCI America
B2329-5G
B2329-25G
TCB2329-5GEA
39.5
CAD
TCB2329-5G
TCB2329-25G
5-Bromo-2-chloropyridine ≥98.0%
5-Bromo-2-chloropyridine
CAS Number: 53939-30-3
MDL Number: MFCD01318951
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70
Other Product Names / Synonyms: 2-Chloro-5-bromopyridine
Formula:
C₅H₃BrClN MW: 192.44 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD01318951 CAS Number: 53939-30-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...