3-Bromo-2-chloropyridine ≥98.0%
Supplier: TCI America
B2328-5G
B2328-25G
TCB2328-5GEA
48.98
CAD
TCB2328-5G
TCB2328-25G
3-Bromo-2-chloropyridine ≥98.0%
3-Bromo-2-chloropyridine
CAS Number: 52200-48-3
MDL Number: MFCD00234007
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 97
Melting point (°C): 57
Other Product Names / Synonyms: 2-Chloro-3-bromopyridine
Formula:
C₅H₃BrClN MW: 192.44 Boiling Pt: 205 Melting Pt: 55 |
MDL Number:
MFCD00234007 CAS Number: 52200-48-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...