2-Bromo-5-nitropyridine ≥98.0%

Supplier: TCI America

B2321-5G B2321-1G
TCB2321-5GEA 105.86 CAD
TCB2321-5G TCB2321-1G
2-Bromo-5-nitropyridine ≥98.0%
2-Bromo-5-nitropyridine

CAS Number: 4487-59-6
Molecular Formula: C5H3BrN2O2
Molecular Weight: 203.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 139
Flash Point (°C): 130

Formula: C₅H₃BrN₂O₂
Boiling Pt: 147 °C
Melting Pt: 139 °C
Flash Pt: 130 °C
MDL Number: MFCD00006222
CAS Number: 4487-59-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3679f98da6c61a703718f4bf152b4b6d326fe4&vid=db3679f98da6c61a703718f4bf152b4b6d326fe4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR