1-Butyl-3-methylimidazoliumhexafluorophosphate ≥98.0%

Supplier: TCI America

Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V), 1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate

B2320-25G B2320-5G
TCB2320-25GEA 137.46 CAD
TCB2320-25G TCB2320-5G
1-Butyl-3-methylimidazoliumhexafluorophosphate ≥98.0%
1-Butyl-3-methylimidazolium hexafluorophosphate

CAS Number: 174501-64-5
MDL Number: MFCD03093295
Molecular Formula: C8H15F6N2P
Molecular Weight: 284.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Clear Liquid
Specific Gravity (20/20): 1.37

Formula: C₈H₁₅F₆N₂P
MW: 284.18 g/mol
Melting Pt: 6
Density: 1.37 (20 °C)
MDL Number: MFCD03093295
CAS Number: 174501-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a83f49974a470bb21566bd00833f3861823901ab&vid=a83f49974a470bb21566bd00833f3861823901ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR