4-(Boc-amino)piperidine 98,0

Supplier: TCI America

Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

B2319-1G B2319-5G
TCB2319-1GEA 28.44 CAD
TCB2319-1G TCB2319-5G
4-(Boc-amino)piperidine 98,0
4-(Boc-amino)piperidine

CAS Number: 73874-95-0
MDL Number: MFCD00798171
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 4-(Boc-amino)piperidine

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00798171
CAS Number: 73874-95-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cd791652ef83ef51aa8d886366196491be85a6e&vid=3cd791652ef83ef51aa8d886366196491be85a6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR