5-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2302-5G B2302-1G
TCB2302-5GEA 115.34 CAD
TCB2302-5G TCB2302-1G
5-Bromo-2-thenoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2-thenoic acid

CAS Number: 7311-63-9
MDL Number: MFCD00079725
Molecular Formula: C5H3BrO2S
Molecular Weight: 207.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 142

Formula: C₅H₃BrO₂S
Melting Pt: 142 °C
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR