4-Biphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

B2294-25G B2294-5G
TCB2294-25GEA 137.46 CAD
TCB2294-25G TCB2294-5G
4-Biphenylboronic acid (contains varying amounts of Anhydride)
4-Biphenylboronic acid

CAS Number: 5122-94-1
Molecular Formula: C12H11BO2
Molecular Weight: 198.03
Form: Crystal
Color: White
Melting point (°C): 266

Formula: C₁₂H₁₁BO₂
Melting Pt: 266 °C
MDL Number: MFCD00093311
CAS Number: 5122-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e870decebae49ec775e334f1a71f22713eaa6987&vid=e870decebae49ec775e334f1a71f22713eaa6987&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR