1,2-Bis(3-indenyl)ethane ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Ethylenebis(1H-indene), 1,2-Di(1H-inden-3-yl)ethane

B2281-1G B2281-5G
TCB2281-1GEA 42.66 CAD
TCB2281-1G TCB2281-5G
1,2-Bis(3-indenyl)ethane ≥98.0%
1,2-Bis(3-indenyl)ethane

CAS Number: 18657-57-3
MDL Number: MFCD00191814
Molecular Formula: C20H18
Molecular Weight: 258.36
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 3,3'-Ethylenebis(1H-indene)

Formula: C₂₀H₁₈
MW: 258.36 g/mol
MDL Number: MFCD00191814
CAS Number: 18657-57-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61f19c6067a1a53967e08f35c9097adb073c0e1f&vid=61f19c6067a1a53967e08f35c9097adb073c0e1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR