N-Benzylglycine ethyl ester ≥96.0%
Supplier: TCI America
B2273-25G
B2273-5G
TCB2273-25GEA
172.22
CAD
TCB2273-25G
TCB2273-5G
N-Benzylglycine ethyl ester ≥96.0%
N-Benzylglycine ethyl ester
CAS Number: 6436-90-4
MDL Number: MFCD00009174
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl (Benzylamino)acetate, Bzl-Gly-OEt
Formula:
C₁₁H₁₅NO₂ Boiling Pt: 142 °C Density: 1.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009174 CAS Number: 6436-90-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...