2-Bromo-4-chloroaniline ≥98.0%
Supplier: TCI America
B2268-25G
B2268-5G
TCB2268-25GEA
167.48
CAD
TCB2268-25G
TCB2268-5G
2-Bromo-4-chloroaniline ≥98.0%
2-Bromo-4-chloroaniline
CAS Number: 873-38-1
Molecular Formula: C6H5BrClN
Molecular Weight: 206.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 66
Formula:
C₆H₅BrClN Boiling Pt: 127 °C Melting Pt: 66 °C |
MDL Number:
MFCD00041313 CAS Number: 873-38-1 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...