N(α)-Boc-D-tryptophan ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Boc-D-Trp-OH

B2259-5G B2259-1G
TCB2259-5GEA 221.52 CAD
TCB2259-5G TCB2259-1G
N(α)-Boc-D-tryptophan ≥98.0% (by HPLC)
N(α)-Boc-D-tryptophan

CAS Number: 5241-64-5
MDL Number: MFCD00037944
Molecular Formula: C16H20N2O4
Molecular Weight: 304.35
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-tryptophan, Boc-D-Trp-OH

Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
MDL Number: MFCD00037944
CAS Number: 5241-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31d6ffe659d7167e55cb10fb9ecd0c2cd1e56cbb&vid=31d6ffe659d7167e55cb10fb9ecd0c2cd1e56cbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR